Learn More
Macherey-Nagel™ Polyamide CC 6
Product Code. 10033091 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Particle Size:
<0.07
0.05 to 0.16 mm
0.10 to 0.3 mm
Unit Size:
1kg
5kg
Product Code. Quantity Particle Size unitSize
10033091 1 kg <0.07 1kg
10328411 1 kg 0.10 to 0.3 mm 1kg
10441862 5 kg 0.05 to 0.16 mm 5kg
10286231 5 kg 0.10 to 0.3 mm 5kg
4 options
Product Code. 10033091
Brand: Macherey-Nagel™ 815610.1