Learn More
Macherey-Nagel™ Polyamide 6 'Chromabond PA Adsorbent
Product Code. 10236521 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity unitSize
10236521 100 g 100g
1 options
Product Code. 10236521
Brand: Macherey-Nagel™ 730660
Specifications
Quantity 100 g
Product Type Chromabond PA Adsorbent
Particle Size 40 to 80 μm
Material Polyamide 6