Learn More
Serva Electrophoresis™ Polyacryl-amide Servalyt Precotes
Product Code. 11447883
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
125 mm
Separation Range:
pH 3 to 10
pH 3 to 6
Unit Size:
Each
Product Code. Length (Metric) Separation Range unitSize
11447883 125 mm pH 3 to 10 Each
11457883 125 mm pH 3 to 6 Each
2 options
Product Code. 11447883
Brand: Serva Electrophoresis™ 4286702
Only null left
Specifications
Gel Percentage 5%
Gel Thickness 0.65 mm
Gel Type IEF
Product Type Homogeneous Page
Separation of Protein
Separation Range pH 3 to 10
Quantity 5/Pk.
Length (Metric) 125 mm
Length (Metric) Gel 125 mm
Width (Metric) Gel 125 mm
compliance-icons
Product Identifier
  • SERVALYT(TM) PRECOTES(TM) Wide Range pH 3-10
Signal Word
  • Danger
Hazard Category
  • Germ cell mutagenicity Category 1
  • Carcinogenicity Category 1
Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .