Learn More
Koflo Mounted In-Line Static Mixers
Product Code. p-8004460 Shop All Cole-Parmer Products
Change view
Click to view available options
Length (Metric):
89 mm
184 mm
286 mm
Unit Size:
Pack of 10
Product Code. Length (Metric) unitSize
11774139 89 mm Pack of 10
11724149 286 mm Pack of 10
11704149 184 mm Pack of 10
3 options
Product Code. 11774139
Brand: Koflo 0466802
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Mounted In-Line Static Mixers can be used with any compression fitting with the same tubing OD.
  • For use with compression fitting with the same tubing OD
Specifications
Diameter (Metric) 8 mm
Length (Metric) 89 mm
Type Mounted In-line Mixer