Learn More
Quickfit™ Polyacetal Joint Clips
Product Code. 11901538
Change view
Click to view available options
Color:
Blue
Brown
Green
Mint
Ochre
Orange
Red
Violet
Yellow
Unit Size:
Pack of 10
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11901538 Green Pack of 10
11991528 Blue Pack of 10
11981528 Yellow Pack of 10
11920955 Red Pack of 10
10796461 Violet Pack of 10
11911538 Orange Pack of 10
10101460 Ochre Pack of 10
10488960 Mint Pack of 10
12029072 Brown Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 11901538 Supplier Quickfit™ Supplier No. KC24
  • ONLINE EXCLUSIVE PRICE
    706.20 NOK
    551.63 NOK / Pack of 10
    Save 154.57 NOK
    21% Off
Estimated Shipment: 11-12-2026
Manufactured from polyacetal for strength and rigidity. Quickfit™ Polyacetal Clamps for Conical Joints are designed to secure corresponding Quickfit™ glassware with conical joints.
Manufactured from polyacetal for strength and rigidity. Quickfit™ Polyacetal Clamps for Conical Joints are designed to secure corresponding Quickfit™ glassware with conical joints.
  • Effortless attachment and removal of clips
  • Reduces the need for traditional clamping
  • Withstands temperatures up to 80°C
  • Color coded sizes for quick identification
Specifications
Quantity 10/Pk.
Type Joint Clamp
Material Polyacetal
For Use With (Equipment) Glassware with conical joints
Size Joint Size 24/29
Color Green