Learn More
Poly(propylene glycol) average Mn 400, BLD Pharm™
Specifications
Specifications
| CAS | 25322-69-4 |
| Molecular Formula | C3nH6n+2On+1 |
| MDL Number | MFCD00084446 |
| Quantity | 500 g |
| SMILES | NONE |
| Chemical Name or Material | Poly(propylene glycol) average Mn 400 |