missing translation for 'onlineSavingsMsg'
Learn More

Poly-L-Lysine Hydrobromide [MW 4,000-15,000], MP Biomedicals

Product Code. 11498340 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
This item is not returnable. View return policy

Product Code. 11498340

Brand: MP Biomedicals 0210269190

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 25988-63-0
Molecular Formula C18H38N6O4
Molecular Weight (g/mol) 402.54
InChI Key WBSCNDJQPKSPII-KKUMJFAQSA-N
Synonym lysyllysyllysine, l-lysyl-l-lysyl-l-lysine, unii-y7f82m80k8, trilysine, h-lys-lys-lys-oh, s-6-amino-2-s-6-amino-2-s-2,6-diaminohexanamido hexanamido hexanoic acid, l-lysine, l-lysyl-l-lysyl, 2olb, lysyl-lysyl-lysine, polylysine hbr
PubChem CID 72363
IUPAC Name (2S)-6-amino-2-[[(2S)-6-amino-2-[[(2S)-2,6-diaminohexanoyl]amino]hexanoyl]amino]hexanoic acid
SMILES C(CCN)CC(C(=O)NC(CCCCN)C(=O)NC(CCCCN)C(=O)O)N

Specifications

Quantity 500 mg
Formula Weight 4,000 to 15,000
Chemical Name or Material Poly-L-Lysine Hydrobromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.