Description
The POLR3E antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human POLR3E. This antibody recognizes human, mouse, rat antigen. The POLR3E antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
Specifications
| Antigen | POLR3E |
| Applications | Western Blot, Immunoprecipitation, ELISA |
| Classification | Polyclonal |
| Concentration | 267 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Quantity | 20 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 55718 |
| Target Species | Human, Rat, Mouse |
| Show More |