Learn More
POLR2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16859904 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16859904 20 μL 20µL
16849904 150 μL 150µL
2 options
Product Code. 16859904
Brand: Proteintech 203701AP20UL
Rabbit Polyclonal Antibody
The antibody is specific to POLR2B.
This gene encodes the second largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. This subunit, in combination with at least two other polymerase subunits, forms a structure within the polymerase that maintains contact in the active site of the enzyme between the DNA template and the newly synthesized RNA.
Specifications
Antigen POLR2B
Applications Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene POLR2B
Gene Accession No. P30876, Q8CFI7
Gene Alias hRPB140, hsRPB2, POL2RB, POLR2B, RNA polymerase II subunit 2, RNA polymerase II subunit B2, RPB2
Gene Symbols Polr2b
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231329, 289561, 5431
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG