missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
POLR1C Polyclonal antibody specifically detects POLR1C in Human samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | POLR1C |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 350 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | AC40, DNA-directed RNA polymerase I subunit C, DNA-directed RNA polymerases I and III 40 kDa polypeptide, POLR1E, polymerase (RNA) I polypeptide C, 30kDa, RNA polymerase I subunit, RPA39RNA polymerases I and III subunit AC1, RPA40TCS3, RPA5RPAC1DNA-directed RNA polymerases I and III subunit RPAC1, RPC40 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human POLR1C (NP_976035.1).,, Sequence:, MAASQAVEEMRSRVVLGEFGVRNVHTTDFPGNYSGYDDAWDQDRFEKNFRVDVVHMDENSLEFDMVGIDAAIANAFRRILLAEVPTMAVEKVLVYNNTSIVQDEILAHRLGLIPIHADPRLFEYRNQGDEEGTEIDTLQFRLQVRCTRNPHAAKDSSDPNELYVNHKVYTRHMTWIPLGNQADLFPEGTIRPVHDDILIA |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?