Learn More
3M™ Polisher ST
Product Code. 17315423 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric):
4.3 cm
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
17315423 4.3 cm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17315423 Supplier 3M™ Supplier No. 7100228760
Estimated Shipment: 28-12-2026
The Polisher ST is a single-use anion exchange (AEX) product intended to reduce host cell protein, viruses, and other negatively charged contaminants in biopharmaceutical process streams.
The 3M™ Polisher ST is intended to reduce host cell protein, viruses, and other negatively charged contaminants in a flow-through polishing chromatography mode in biopharmaceutical process streams. The synthetic, hybrid purifier contains two complementary AEX-functional media: a quaternary ammonium ('Q') functional nonwoven and a guanidinium-functional membrane.
The Q-functional nonwoven provides reduction of turbidity (when present) and DNA. It also provides a portion of the product's AEX capacity for HCP and virus reduction. Guanidinium functionalized membrane downstream of the functionalized nonwoven provides additional AEX polishing capacity. The novel guanidinium functionality of the downstream AEX polishing membrane mimics the amino acid, arginine, one of the three positively charged, naturally occurring standard amino acids that make up proteins and is responsible for viral clearance and high salt tolerance.
Specifications
Disposable Single-use
Material Polypropylene
Material (Membrane) Polyamide
Filtration Area 1 cm2
Flow Rate 1 mL/min.
Inlet Style Luer Lock
No. per Case 4
Outlet Style Luer Lock
Pressure Rating 1.4 bar
Temperature (English) Operating 104°F
Temperature (Metric) Operating 40°C
Height (English) 1.9 in.
Height (Metric) 4.8 cm
Diameter (English) 1.7 in.
Diameter (Metric) 4.3 cm