missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ POLH (Human) Recombinant Protein
Human POLH full-length ORF ( AAH15742, 1 a.a. - 414 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005429-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MATGQDRVVALVDMDCFFVQVEQRQNPHLRNKPCAVVQYKSWKGGGIIAVSYEARAFGVTRSMWADDAKKLCPDLLLAQVRESRGKANLTKYREASVEVMEIMSRFAVIERASIDEAYVDLTSAVQERLQKLQGQPISADLLPSTYIEGLPQGPTTAEETVQKEGMRKQGLFQWLDSLQIDNLTSPDLQLTVGAVIVEEMRAAIERETGFQCSAGISHNKVLAKLACGLNKPNRQTLVSHGSVPQLFSQMPIRKIRSLGGKLGASVIEILGIEYMGELTQFTESQLQSHFGEKNGSWLYAMCRGIEHDPVKPRQLPKTIGCSKNFPGKTALATREQVQWWLLQLAQELEERLTKDRNDNDRVATQLVVSIRVQGDKRLSSLRRCCALTRYDAHKMSHDAFTVIKNCNTSGIQTE
Specifications
AAH15742 | |
polymerase (DNA directed), eta | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
POLH | |
GST |
5429 | |
Wheat germ expression system | |
10 μg | |
FLJ16395, FLJ21978, RAD30, RAD30A, XP-V, XPV | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |