missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLD2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29765-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
POLD2 Polyclonal antibody apecifically detetects POLD2 antigen in Human Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.Specifications
POLD2 | |
Polyclonal | |
Unconjugated | |
BC000459 | |
Rabbit | |
Antigen affinity purification | |
Primary | |
Human | |
Liquid |
Western Blot, Immunohistochemistry, ELISA | |
400 μg/mL | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
POLD2 | |
POLD2 fusion protein Ag31521 | |
150 μL | |
5425 | |
Store at -20°C. Stable for one year after shipment. | |
IgG |