Learn More
Cole-Parmer™ PolarSafe™ Dewar and Traceable™ Temperature Data Logger Bundles
Shop All Cole-Parmer Products
Click to view available options
Capacity (Metric):
2 L
30 L
35 L
47 L
95 L
115 L
Unit Size:
Each
Description
- Improve sample protection by maintaining specific temperature ranges for extended time periods
- Maintain sample integrity with Traceable™ temperature monitoring
- Data logger is provided with a 2-year NIST-traceable calibration certificate
Specifications
Specifications
| Capacity (Metric) | 30 L |
| Depth (English) | 25.88 in. |
| Depth (Metric) | 65.72 cm |
| Diameter (English) Inner | 4.88 in. |
| Diameter (English) Outer | 18.9 in. |
| Diameter (Metric) Inner | 12.5 cm |
| Diameter (Metric) Outer | 48.01 cm |
| Material | Aluminum |