missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Pokeweed mitogen native Protein (Crude) from pokeweeds

Product Code. 16292270
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 16292270

Brand: Abnova™ P5356.5mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Enzyme, SDS-PAGE

Specifications

For Use With (Application) Enzyme Activity, SDS-PAGE
Name Mitogen (Pokeweed) Native Protein
Preparation Method Native protein purified from pokeweeds
Quantity 5 mg
Storage Requirements Store at -20°C on dry atmosphere.. After reconstitution with distilled water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name MITOGEN
Biological Activity Shows agglutination when tested with human red blood cells.
Species Plants
Protein Tag None
Expression System Native protein purified from pokeweeds
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.