Learn More
Podoplanin Mouse anti-Human, Unconjugated, Clone: 1D9F3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67432-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16800066
  • 1611.84 NOK / 20µL
Estimated Shipment: 24-05-2024
to see stock.
Description
Description
The Podoplanin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Podoplanin. This antibody recognizes human antigen. The Podoplanin antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
Podoplanin
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Immunohistochemistry, ELISA
1D9F3
Unconjugated
20 μL
10630
IgG2a