missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PNMA1 encodes a protein that is highly restricted to the brain and testis. Anti-PNMA1 reacts mainly with subnuclear elements (including the nucleoli) and to a lesser degree the cytoplasm.
Specifications
Specifications
| Antigen | PNMA1 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PNMA1 |
| Gene Accession No. | Q8C1C8, Q8ND90, Q8VHZ4 |
| Gene Alias | 37 kDa neuronal protein, MA1, paraneoplastic antigen MA1, PNMA1 |
| Gene Symbols | Pnma1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?