Learn More
Invitrogen™ Pneumolysin Polyclonal Antibody
Description
Pneumolysin Polyclonal Antibody for Western Blot, ELISA
Pneumolysin (pneumococcal hemolysin) is an intracellular protein produced by all strains of S. pneumoniae and is a cytoplasmic cholesterol-dependent cytolysin forming large pores in cholesterol-containing membranes and are therefore cytotoxic to mammalian cells. The molecular weight of Pneumolysin is approximately 53 kDa.
Specifications
Specifications
| Antigen | Pneumolysin |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 |
| Gene | ply |
| Gene Accession No. | Q04IN8 |
| Gene Alias | ply; Pneumolysin (Thiol-activated cytolysin) |
| Gene Symbols | ply |
| Show More |