missing translation for 'onlineSavingsMsg'
Learn More

PNCK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13712-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16892064

  • 3468.15 NOK / 150µL
Estimated Shipment: 15-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PNCK is a member of the calcium/calmodulin-dependent protein kinase family of protein serine/threonine kinases (Gardner et al., 2000 ).
Specifications

Specifications

PNCK
Polyclonal
Unconjugated
PNCK
BSTK3, CaM KI beta, CaM kinase I beta, CaM kinase IB, CaMK1b, CaMKI beta, PNCK
Rabbit
Antigen Affinity Chromatography
RUO
139728, 29660, 93843
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O70150, Q6P2M8, Q9QYK9
Pnck
PNCK Fusion Protein Ag4650
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers