Learn More
VITLAB™ PMP Griffin Beaker
Product Code. 10109370
Change view
Click to view available options
Capacity (Metric):
25 mL
100 mL
150 mL
400 mL
500 mL
600 mL
1000 mL
2000 mL
3000 mL
5000 mL
Unit Size:
Each
Pack of 12
Pack of 6
Product Code. Capacity (Metric) unitSize
10109370 1000 mL Each
12783399 25 mL Pack of 12
12703409 100 mL Pack of 12
10722542 100 mL Each
10210891 150 mL Each
10732542 400 mL Each
12723409 500 mL Pack of 6
10201131 600 mL Each
12733409 1000 mL Pack of 6
10510012 2000 mL Each
10189410 3000 mL Each
10373141 5000 mL Each
12 options
Product Code. 10109370
Brand: VITLAB™ 61403
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Make your job easier and safer while handling a wide range of liquids with these polymethylpentene beakers. VITLAB™ PMP Griffin Beakers feature an easy to read, embossed red scale.
  • Crystal clear transparent graduated beakers
  • For autoclaving, models with raised graduations are recommended
  • To preserve markings, clean at no higher than 60°C
Specifications
Color Crystal Clear
Capacity (Metric) 1000 mL
Material PMP
For Use With (Application) For Volume Measurement
Height (Metric) 147 mm
Type Griffin Beaker
Autoclavable Autoclavable
Graduated Yes
Certifications/Compliance ISO 7056
Scale Red Printed Scale
Style Griffin