Learn More
BRAND™ Rack for Sedimentation Cones
Holds 2 Imhoff sedimentation cones made of plastic. Compact design and easy to carry, even with cones filled.
Brand: BRAND™ 388050
1277.75 NOK valid until 2024-04-26
Use promo code "21699" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
- Holds 2 Imhoff sedimentation cones
- Polymethyl methacrylate (PMMA)/Polypropylene (PP) construction
Specifications
Imhoff Cone Rack | |
PMMA, Polypropylene | |
300 x 130 x 315mm |