missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PML Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21041-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is one of two human homologs of Saccharomyces cerevisiae Rad23, a protein involved in nucleotide excision repair (NER). This protein was shown to interact with, and elevate the nucleotide excision activity of 3-methyladenine-DNA glycosylase (MPG), which suggested a role in DNA damage recognition in base excision repair. This protein contains an N-terminal ubiquitin-like domain, which was reported to interact with 26S proteasome, as well as with ubiquitin protein ligase E6AP, and thus suggests that this protein may be involved in the ubiquitin mediated proteolytic pathway in cells.Specifications
PML | |
Polyclonal | |
Unconjugated | |
PML | |
MYL, PML, PP8675, promyelocytic leukemia, RING finger protein 71, RNF71, TRIM19 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5371 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P29590 | |
PML | |
PML Fusion Protein Ag15377 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |