missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMEL17/SILV Antibody (Novartis patent anti-PMEL17) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28130JF669
This item is not returnable.
View return policy
Description
PMEL17/SILV Monoclonal antibody specifically detects PMEL17/SILV in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| PMEL17/SILV | |
| Monoclonal | |
| Janelia Fluor 669 | |
| D12S53EP1, gp100, ME20, ME20-M, melanocyte protein mel 17, Melanocyte protein Pmel 17, Melanocytes lineage-specific antigen GP100, Melanoma-associated ME20 antigen, melanosomal matrix protein17, PMEL17P100, premelanosome proteinME20M, SI, SIL, silver (mouse homolog) like, silver homolog (mouse), Silver locus protein homolog, silver, mouse, homolog of, SILVPmel17 | |
| PMEL | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Novartis patent anti-PMEL17 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| 6490 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction