missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLXNA4 Monoclonal Antibody (707206)
GREENER_CHOICE

Product Code. 15594012
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15594012 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15594012 Supplier Invitrogen™ Supplier No. MA524326

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

In direct ELISAs, no cross-reactivity with recombinant human Plexin A1, recombinant mouse (rm) Plexin A1 or rmPlexin A2 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

PLXNA4 is a protein coding gene. Coreceptor for SEMA3A. Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance in the developing nervous system. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin. The plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLXNA4
Applications Flow Cytometry
Classification Monoclonal
Clone 707206
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative; pH 7.4
Gene Plxna4
Gene Accession No. Q9HCM2
Gene Alias 9330117B14; FAYV2820; Kiaa1550; mKIAA1550; OTTHUMP00000207245; PLEXA4; plexin A4; plexin A4, A; plexin-A4; Plxa4; Plxna4; Plxna4a; PLXNA4B; PRO34003; UNQ2820/PRO34003
Gene Symbols Plxna4
Host Species Mouse
Immunogen CHO-derived recombinant human Plexin A4 Thr24-Pro1237.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91584
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.