Learn More
Emma Safety Footwear Pluvius Safety Shoes
Product Code. 13419599
Change view
Click to view available options
Size:
41
42
43
44
46
49
Unit Size:
1 pair
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
13419599 41 1 pair
13439599 42 1 pair
13459599 43 1 pair
13469599 44 1 pair
13489599 46 1 pair
15979091 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 13419599 Supplier Emma Safety Footwear Supplier No. MM93786841
  • ONLINE EXCLUSIVE PRICE
    2625.00 NOK
    2096.95 NOK / 1 pairs
    Save 528.05 NOK
    20% Off
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
Protected with Sympatex™ one of the hardest wearing, most liquid repellant materials available, including corrosive chemicals. Provides outstanding grip in every kind of weather and a good on-foot fit.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather / Sympatex™ construction
  • Chemical resistant / Liquid repellant
  • GripForce™ Duo Heat Resistant Outsole
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black, Gray
Design Pluvius
Liner Material Hydro-Tec™ Sanitized Silver / Sympatex™
Material Leather
Size 41
Toe Material Steel
Resistant To Chemicals