Learn More
Memmert™ Plug-In Tube Extension for SN450, Straight
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
Brand:  Memmert™ SN450B29718
Additional Details : Weight : 0.01200kg
Product Code. 17568625
  • 510.00 NOK / Each
Estimated Shipment: 07-06-2024
Description
  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight
Specifications
Specifications
60.3 mm
SN450 Sterilizer