missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PLSCR1 (Human) Recombinant Protein
Human PLSCR1 full-length ORF ( AAH32718, 1 a.a. - 318 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005359-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MDKQNSQMNASHPETNLPVGYPPQYPPTAFQGPPGYSGYPGPQVSYPPPPAGHSGPGPAGFPVPNQPVYNQPVYNQPVGAAGVPWMPAPQPPLNCPPGLEYLSQIDQILIHQQIKLLEVLTGFETNNKYEIKNSFGQRVYFAAEDTDCCTRNCCGPSRPFTLRIIDNMGQEVITLERPLRCSSCCCPCCLQEIEIQAPPGVPIGYVIQTWHPCLPKFTIQNEKREDVLKISGPCVVCSCCGDVDFEIKSLDEQCVVGKISKHWTGILREAFTDADNFGIQFPLDLDVKMKAVMIGACFLIDFMFFESTGSQEQKSGVW
Specifications
AAH32718 | |
phospholipid scramblase 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PLSCR1 | |
GST |
5359 | |
Wheat germ expression system | |
25 μg | |
MMTRA1B | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |