missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLRG1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11914-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Plrg1 is a protein coding gene involved in pre-mRNA splicing as component of the spliceosome. Component of the PRP19-CDC5L complex that forms an integral part of the spliceosome and is required for activating pre-mRNA splicing.Specifications
PLRG1 | |
Polyclonal | |
Unconjugated | |
PLRG1 | |
Pleiotropic regulator 1, PLRG1, PRL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5356 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43660 | |
PLRG1 | |
PLRG1 Fusion Protein Ag2527 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |