Learn More
PLP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21482-1-AP-150UL
This item is not returnable.
View return policy
Description
PLP2 may play a role in cell differentiation in the intestinal epithelium.Specifications
| PLP2 | |
| Polyclonal | |
| Unconjugated | |
| PLP2 | |
| A4, A4 LSB, Intestinal membrane A4 protein, PLP2, Proteolipid protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5355 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q04941 | |
| PLP2 | |
| PLP2 Fusion Protein Ag15740 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |