missing translation for 'onlineSavingsMsg'
Learn More

PLP2 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16851075 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16851075 20 μL 20µL
16841075 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16851075

Brand: Proteintech 214821AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PLP2 may play a role in cell differentiation in the intestinal epithelium.

Specifications

Antigen PLP2
Applications Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PLP2
Gene Accession No. Q04941
Gene Alias A4, A4 LSB, Intestinal membrane A4 protein, PLP2, Proteolipid protein 2
Gene Symbols PLP2
Host Species Rabbit
Immunogen PLP2 Fusion Protein Ag15740
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5355
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.