Learn More
Hecht Karl™ Plexiglas Test Tube Racks
Product Code. p-8004448
Click to view available options
Height (Metric):
75 mm
76 mm
79 mm
Unit Size:
Each
Product Code. 10503652
Brand: Hecht Karl™ 42807010
Only null left
Specifications
Product Type Test Tube Rack
Holds Tubes (13 mm)
Material Plexiglas
Positions 12
Quantity 1 Pack
Height (Metric) 76 mm
Diameter (Metric) Opening 13 mm