Learn More
Hecht Karl™ Plexiglas Test Tube Racks
Specifications
Specifications
| Holds | Tubes (18 mm) |
| Material | Plexiglas |
| Positions | 12 |
| Quantity | 1 Pack |
| Diameter (Metric) Opening | 18 mm |
| Product Type | Test Tube Rack |
| Height (Metric) | 75 mm |