missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLCD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14228-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Phosphoinositide-specific phospholipase C (PLC) acts as a signal transducer that generates 2 second messengers, diacylglycerol and inositol 1, 4, 5-trisphosphate, by hydrolyzing inositol phospholipids. PLC comprises a diverse family of enzymes that differ in structure and tissue distribution (Berridge, 1993 ).Specifications
PLCD1 | |
Polyclonal | |
Unconjugated | |
PLCD1 | |
Phospholipase C delta 1, Phospholipase C III, phospholipase C, delta 1, PLC delta 1, PLC III, PLCD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5333 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P51178 | |
PLCD1 | |
PLCD1 Fusion Protein Ag5468 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |