missing translation for 'onlineSavingsMsg'
Learn More

PLCD1 Mouse anti-Human, Unconjugated, Clone: 1D10B11, Proteintech

Product Code. 16899465 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16899465 150 μL 150µL
16809475 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16899465

Brand: Proteintech 670641IG150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The PLCD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PLCD1. This antibody recognizes human antigen. The PLCD1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Antigen PLCD1
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
Clone 1D10B11
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 5333
Target Species Human
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.