missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PLCB2 (Human) Recombinant Protein

Human PLCB2 full-length ORF ( AAH00939, 1 a.a. - 199 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00005330-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16139171

  • 6270.00 NOK / 25µg
Estimated Shipment: 25-11-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MSLLNPVLLPPKVKAYLSQGERFIKWDDETTVASPVILRVDPKGYYLYWTYQSKEMEFLDITSIRDTRFGKFAKMPKSQKLRDVFNMDFPDNSFLLKTLTVVSGPDMVDLTFHNFVPYKENVGKAWAEDVLALVKHPLTANASRSTFLDKILVKLKMQLNSEGKIPVKNFFQMFPADRKRVEAALSACHLPKGKPGGAR
Specifications

Specifications

AAH00939
phospholipase C, beta 2
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PLCB2
GST
5330
Wheat germ expression system
25 μg
FLJ38135
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates
Special Offers

Special Offers