Learn More
Platinum rod, 6.35mm (0.25in) dia, Premion™, 99.99% (metals basis)
Product Code. 11397397
Change view
Click to view available options
Quantity:
50 mm
Unit Size:
50mm
Product Code. Quantity unitSize
11397397 50 mm 50mm
1 options
Product Code. 11397397
Brand: Thermo Scientific Alfa Aesar 012556.DJ
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 7440-06-4
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 6-4-7440
Quantity 50 mm
Percent Purity 99.99%