Learn More
Platinum Iridium foil, 0.025mm (0.001in) thick, 99.9% (metals basis)
Product Code. 11354757
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Unit Size:
25mm
Each
Product Code. Quantity unitSize
11354757 25 x 25 mm 25mm
11375929 50 x 50 mm Each
11348968 100 x 100 mm Each
3 options
Product Code. 11354757
Brand: Thermo Scientific Alfa Aesar 041802.FF
Request Bulk or Custom Format
Used in the fabrication of micro electrodes for electrical stimulation of nervous tissue and electrophysiological recordings. | Pt:Ir; 90:10 wt%
Platinum Iridium foil, 0.025mm (0.001in) thick, 99.9% (metals basis) is used in the fabrication of micro electrodes for electrical stimulation of nervous tissue and electrophysiological recordings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Platinum Iridium foil
Quantity 25 x 25 mm
Molecular Formula Pt:Ir; 90:10 wt%
MDL Number MFCD02091683
Solubility Information Insoluble in water.
Percent Purity 99.9%
Packaging Tube