Learn More
Dostmann Electronic Platinum Immersion Probe
Product Code. 10701654
Change view
Click to view available options
Length (Metric):
150 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
10701654 150 mm Each
1 options
Product Code. 10701654
Brand: Dostmann Electronic 60001073
Only null left
Add to basket
Add to basket
With a response time of 12 seconds and a temperature range of –200 to 500°, this platinum immersion probe is ideal for many research, scientific, and laboratory environments.
  • Temperature Probe Pt100 6000-1073
  • High-accuracy resistance probe, Pt100 1/10-DIN, Platinium-ceramic sensor IEC 751, 4-wire, cable 1000mm PVC
  • recommend for accurate measurings e.g. with certificate
  • Immersion probe, with handle, mineral-insulated, WS 1.4571, -200..+450°C, 150 x 3.0mm
Specifications
Response Time 12 sec.
Temperature Range (Metric) -200°C to +500°C
Length (Metric) 150 mm