Learn More
ÖGUSSA Platinum Evaporating Basin with Flat Bottom
Click to view available options
Capacity (Metric):
20 mL
50 mL
90 mL
150 mL
Unit Size:
Each
Description
- Flat bottom
- Platinum
- Heat resistant
Specifications
Specifications
| Bottom Shape | Flat |
| Capacity (Metric) | 20 mL |
| Material | Platinum |
| Type | Evaporating Basin |
| Height (Metric) | 20 mm |
| Diameter (Metric) | 40 mm |