Learn More
ÖGUSSA Platinum Evaporating Basin with Flat Bottom
Description
- Flat bottom
- Platinum
- Heat resistant
Specifications
Specifications
| Bottom Shape | Flat |
| Capacity (Metric) | 90 mL |
| Material | Platinum |
| Type | Evaporating Basin |
| Height (Metric) | 35 mm |
| Diameter (Metric) | 70 mm |