Learn More
Eppendorf™ Platforms for Innova™ 2150 Shaker
Description
- Holds Erlenmeyer flasks
- Dedicated platform
- Made of phenolic resin
Specifications
Specifications
| Holds | 500 mL Erlenmeyer Flasks |
| Material | Phenolic Resin |
| Dimensions (L x W) | 61 x 45.7 cm |
| For Use With (Equipment) | Innova™ 2300/43/43R and Excella™ E10/E25/E25R Shakers |
| No. of Positions | 20 |