Learn More
Grant Instruments™ Platform for Grant Orbital Shakers and Shaker Incubators
Description
- Includes clamps to keep vessels secure
- For use with
- ES-80 Shaker Incubator
- PSU 20i Orbital Shaker
Specifications
Specifications
| Holds | 16 x 250 to 300 mL Flasks |
| For Use With (Equipment) | PSU-20i Orbital Platform Shaker |
| No. of Positions | 16 |