Learn More
Kinematica™ ST-P13/320 Holder System for PT 1200 E
Product Code. 11985089 Shop All Kinematica Products
Click to view available options
For Use With (Equipment):
Polytron™ PT 1200 E
Unit Size:
Each
Product Code. 11985089
Brand: Kinematica™ 11040007
Stainless steel plate stand with anti-slip mat made of silicone, with height-adjustable holder for the drive and for fastening the base station.
  • Stainless steel plate stand with anti-slip silicone mat
  • Drive unit height adjustable allowing for hands free work
  • Vessel Holder
  • Safety Positioning ring: safety feature to prevent uncontrolled adjustment of drive unit
Specifications
Product Type Plate Stand
Description Stand for PT 1200E with height-adjustable holder for the drive unit and fastening the base station.
For Use With (Equipment) Polytron™ PT 1200 E
Includes Silicone Anti-Slip Mat
Model ST-P 13/320