Learn More
Thermo Scientific™ Plate Count Agar
Product Code. p-4523948
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
Media Type:
Plate Count Agar
Manufacturing Origin:
Germany
United Kingdom
Unit Size:
Pack of 10
Product Code. 11973612
Brand: Thermo Scientific™ PO0158A
Enumerate viable organisms in milk, water, food and dairy products using this non-selective medium.
Enumerate viable organisms in milk, dairy products, water, food and other specimens of sanitary importance using non-selective Thermo Scientific™ Plate Count Agar.
This standard medium meets the formulation of APHA, AOAC and PHLS.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Tested in accordance with CEN ISO/TS 11133-2.
Color Straw, clear
Content And Storage 2° to 10°C, away from light
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Media Type Plate Count Agar
Product Type Agar
Fill Weight 19.5g ± 1.0g
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.