missing translation for 'onlineSavingsMsg'
Learn More

Scharlab Plate Count Agar (PCA)

Product Code. 11451847
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451847 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11451847 Supplier Scharlab Supplier No. 01161500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Scharlab Plate Count Agar is for use with aerobic plate counts by the surface inoculation method.

  • BAM/COMPF/IDF/IFU/ISO/USP
  • Medium for aerobic plate counts by the surface inoculation method (standard Plate Count Agar)
  • According to ISO 4833, 8552, 17410 and IFU No. 6 standards

Specifications

Certifications/Compliance ISO 4833, 8552, 17410
Form Solid
Media Type Plate Count Agar
Product Type Microbiology Media
Formulation Casein Peptone 5.0 g/L, Yeast Extract 2.5 g/L, Dextrose 1.0 g/L, Agar 15.0 g/L
For Use With (Application) Aerobic Plate Counts
Incubation Conditions 30°C ± 1°C
Incubation Time 72 ±3 hr.
pH 7.0 ±0.2
Quantity 500 g
Solubility Insoluble
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.