Learn More
BD PlastiPak™ Luer Slip Syringe without Needle
Description
- Sterile disposable
- Polypropylene
- Lubricant: Silicone (<25μg/sq. mm)
- Permanent ink markings
- Polypropylene cap
Certifications
EN ISO 13485
Specifications
Specifications
| Product Type | Central Cone |
| Graduations | 0.01 mL |
| Volume (Metric) | 1 mL |
| Sterility | Sterile |