Learn More
BD PlastiPak™ Syringe with Luer Lock
Product Code. 15544835
Change view
Click to view available options
Volume (Metric):
10 mL
Graduations:
0.2 mL
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Graduations Unit Size
15544835 10 mL 0.2 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15544835 Supplier BD Supplier No. 305959
  • ONLINE EXCLUSIVE PRICE
    1150.00 NOK
    629.67 NOK / Pack of 100
    Save 520.33 NOK
    45% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Used for injections and sampling.
  • 10mL capacity with 0.2mL graduations
  • Sterile and disposable
  • Polypropylene construct
  • Clear barrel for perfect visibility of syringe contents
  • Special silicone lubrication ensures that plunger moves smoothly and evenly
  • Sterilized by ethylene oxide
Certifications
EN ISO 7886-1
TRUSTED_SUSTAINABILITY
Specifications
Graduations 0.2 mL
Volume (Metric) 10 mL