Learn More
Kartell™ Plastilab™ Untapered Y-Shaped Tubing Connectors
Designed for use with hose system and vacuum lines.
Brand: Kartell™ 7046600
Additional Details : Weight : 0.01440kg
Description
- Made from polypropylene
- Autoclavable
- Nominal dimensions
Compliance
Meet Food and Agriculture regulations
Specifications
Untapered Y Tubing Connector | |
8 mm | |
Polypropylene | |
10/Pk. | |
Y |
Meet with the Food and Drug regulations. 90/128/CEE 92/39/CEE e FDA 21 CFR 177.1520, CE and IVD | |
Autoclavable | |
Useful for any hoses system and vacuum line, suitable for food stuff | |
5.5mm bore |