Learn More
Kartell™ Plastilab™ Polypropylene 4-Way Connectors
For use with plastic or rubber tubing.
Brand: Kartell™ 0141600
Additional Details : Weight : 0.10000kg
Description
- Sturdy shape provides strength
- Chemical and thermal resistant
Specifications
CE IVD | |
Autoclavable | |
100/Pk. | |
4-way cross |
15 mm | |
Polypropylene | |
11.6mm Bore | |
Natural |