Learn More
Kartell™ Plastilab™ Polypropylene 4-Way Connectors
Product Code. 12569756
Change view
Click to view available options
Port Size:
11.6mm Bore
4.6mm Bore
Packaging Quantity:
10/Pk.
100/Pk.
Unit Size:
Pack of 10
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Port Size Packaging Quantity Unit Size
12569756 4.6mm Bore 10/Pk. Pack of 10
15598265 11.6mm Bore 100/Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12569756 Supplier Kartell™ Supplier No. 7141200
  • ONLINE EXCLUSIVE PRICE
    164.00 NOK
    144.49 NOK / Pack of 10
    Save 19.51 NOK
    11% Off
In Stock
Add to basket
Edge
Add to basket
For use with plastic or rubber tubing.
  • Sturdy shape provides strength
  • Chemical and thermal resistant
Specifications
Certifications/Compliance CE IVD
Diameter (Metric) 8 mm
Autoclavable Autoclavable
Material Polypropylene
Packaging Quantity 10/Pk.
Port Size 4.6mm Bore
Shape 4-way cross
Color Natural